missing translation for 'onlineSavingsMsg'
Learn More

Erythromycin USP, Medisca™

$113.46 - $1358.60

Chemical Identifiers

CAS 114-07-8
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
18-048-035
View Documents
Medisca Pharmaceutique Inc
084205
100 g
Each for $340.42
Only null left
 
18-048-049
View Documents
Medisca Pharmaceutique Inc
084208
500 g
Each for $1,358.60
Only null left
 
18-048-112
View Documents
Medisca Pharmaceutique Inc
084204
25 g
Each for $113.46
Only null left
 
Description

Description

Specifications

Chemical Identifiers

114-07-8

Specifications

α-Carboxybenzyl Penicillin Disodium Salt
114-07-8
100 g
USP
Crystalline Powder
White or White/Yellow
85.0 to 100.5%
Videos
SDS
Documents

Documents