accessibility menu, dialog, popup

UserName

CAS RN 114-07-8

IUPAC Name: (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-{[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-14-ethyl-7,12,13-trihydroxy-4-{[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy}-3,5,7,9,11,13-hexamethyl-1-oxacyclotetradecane-2,10-dione
Synonyms: erythromycin erytop erythromycin phosphate (substance) erythrogran erysafe erytab eryacne erymed erymax derimer
Molecular Weight (g/mol): 733.94
Molecular Formula: C37H67NO13
InChi Key: ULGZDMOVFRHVEP-RWJQBGPGSA-N
SMILES: CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O
Color 
  • (2)
  • (2)
  • (3)
  • (2)
Physical Form 
  • (7)
  • (2)
Quantity 
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
Type 
  • (3)
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (7)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)