missing translation for 'onlineSavingsMsg'
Learn More

Erythromycin USP, Medisca™
GREENER_CHOICE

Catalog No. 18048035
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18048035 100 g
18048049 500 g
18048112 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18048035 Supplier Medisca Pharmaceutique Inc Supplier No. 084205

Chemical Identifiers

CAS 114-07-8

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Crystalline Powder
Color White or White/Yellow
Quantity 100 g
Percent Purity 85.0 to 100.5%
Grade USP