missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erythromycin Ethylsuccinate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 1264-62-6 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C43H75 NO16 |
| MDL Number | MFCD00084692 |
| Synonym | Erythromycin ethyl succinate |
| Type | Certified Reference Material |
| Quantity | 250 mg |
| Formula Weight | 862.05 |
| Grade | Certified Reference Material |