missing translation for 'onlineSavingsMsg'
Learn More

Erythritol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111002318
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002318 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002318 Supplier Merck Emd Millipore Supplier No. PHR14791G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Featured Industries: Pharmaceutical (small molecule)

Specifications

Packaging Ampule
CAS 149-32-6
Linear Formula HOCH2 [CH(OH)]2 CH2 OH
MDL Number MFCD00004710
Melting Point 118°C to 120°C
Boiling Point 329°C to 331°C
Synonym meso-Erythritol; 1,2,3,4-Butanetetrol; meso-1,2,3,4-Tetrahydroxybutane; i-Erythritol
Type Certified Reference Material
Quantity 1 g
Formula Weight 122.12
Grade Certified Reference Material
Show More Show Less