missing translation for 'onlineSavingsMsg'
Learn More

Erioglaucine sodium salt

Catalog No. AAJ6178014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6178014 25 g
AAJ6178006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6178014 Supplier Thermo Scientific Chemicals Supplier No. J6178014

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3844-45-9 | C37H34N2Na2O9S3 | 792.844 g/mol

Erioglaucine sodium salt is used as a colorant for foods and other substances to induce a color change.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Erioglaucine sodium salt is used as a colorant for foods and other substances to induce a color change.

Solubility
Soluble in water (30 mg/ml), ethanol.

Notes
Dye content: approx 60%

Chemical Identifiers

CAS 3844-45-9
Molecular Formula C37H34N2Na2O9S3
Molecular Weight (g/mol) 792.844
MDL Number MFCD00012141
InChI Key SGHZXLIDFTYFHQ-UHFFFAOYSA-L
Synonym C.I. 42090; Acid Blue 9
PubChem CID 19700
ChEBI CHEBI:82411
IUPAC Name disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzenesulfonate
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=CC=C5S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 283°C
Merck Index 14,1373
Color Purple
Solubility Information Soluble in water (30mg/ml),ethanol.
Quantity 25 g
Physical Form Powder
Formula Weight 792.86
Chemical Name or Material Erioglaucine sodium salt
Hazard Category H500
EINECSNumber 223-339-8
RTECSNumber BQ4725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only