missing translation for 'onlineSavingsMsg'
Learn More

Ergosterol 95.0+%, TCI America™

Catalog No. E00185G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E00185G 5 g
E001825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E00185G Supplier TCI America Supplier No. E00185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57-87-4
Molecular Formula C28H44O
Molecular Weight (g/mol) 396.659
MDL Number MFCD00003623
InChI Key DNVPQKQSNYMLRS-APGDWVJJSA-N
Synonym ergosterol, provitamin d2, unii-z30ray509f, ergosterin, ergosta-5,7,22e-trien-3beta-ol, 5,7,22-ergostatrien-3beta-ol, 3beta-hydroxy-5,7,22-ergostatriene, 22e,24s-24-methylcholesta-5,7,22-trien-3beta-ol, 22e-ergosta-5,7,22-trien-3beta-ol, ergosta-5,7,22-trien-3-ol, 3b,22e
PubChem CID 444679
ChEBI CHEBI:16933
IUPAC Name (3S,9S,10R,13R,14R,17R)-17-[(E,2R,5R)-5,6-dimethylhept-3-en-2-yl]-10,13-dimethyl-2,3,4,9,11,12,14,15,16,17-decahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC(C)C(C)C=CC(C)C1CCC2C1(CCC3C2=CC=C4C3(CCC(C4)O)C)C

Specifications

Melting Point 159°C
Color White-Yellow
Boiling Point 250°C
Quantity 5 g
Formula Weight 396.66
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ergosterol
EINECSNumber (9)-0248
TSCA No
Recommended Storage Refrigerator