missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Ergomate Pipet Accessories

Catalog No. 14559560
Change view
Click to view available options
Volume (Metric):
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14559560 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14559560 Supplier Sartorius Supplier No. 783500

Accessory for Ergomate Pipet

Ergomate removes the need to hold the plate at any angle and allows safe dispensing, resulting easier, safer and more precise pipetting.

According to ISO 8655 recommendations, dispensing especially small volumes should be done against the inside wall of receiving vessel. Ergomate is an ideal tool for finding proper positioning in pipetting.

Microplate grids for quick identification and sample management.

Grids are double sided. Reverse side featuring numbering from 1-12.

Plate Holder

When reversed plate holder creates an ideal aerosol barrier for use in hoods or on bench top surfaces.

Plate holder can be reversed and used for flat surface pipetting or as an incubation station with lid.

Ideal for safe transporting of plates.

Non autoclavable

Specifications

Autoclavable Autoclavable
For Use With Pipettes
Material Polypropylene
Type 120mL reagent vessel
Volume (Metric) 120 mL
For Use With (Equipment) Multichannel Pipettes
Quantity 120mL