missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERCC3 Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine and Orangutan
The human XPB DNA helicase is a subunit of the DNA repair/basal transcription factor TFIIH, and is involved in early steps of the nucleotide excision repair (NER) pathway. Two distinct clinical phenotypes, xeroderma pigmentosum associated with Cockayne′s syndrome (XP/CS) and trichothiodystrophy (TTD), can be due to mutations in the XPB gene.
Spécifications
Spécifications
| Antigène | ERCC3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Formule | TBS with 0.1% BSA and 0.09% sodium azide |
| Gène | ERCC3 |
| Numéro d’acquisition du gène | P19447, P49135 |
| Alias du gène | BTF2, GTF2H, RAD25, TFIIH, XPB |
| Symboles de gène | ERCC3 |
| Espèces hôtes | Rabbit |
| Afficher plus de résultats |