missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) nitrate pentahydrate, 99.9%, (trace metal basis)

Catalog No. AC315141000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC315141000 100 g Plastic bottle
AC315140250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC315141000 Supplier Thermo Scientific Chemicals Supplier No. 315141000

missing translation for 'validMainframeMsg'

CAS: 10031-51-3 | ErH10N3O14 | 443.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 10031-51-3
Molecular Formula ErH10N3O14
Molecular Weight (g/mol) 443.35
MDL Number MFCD00149691
InChI Key LWHHUEHWVBVASY-UHFFFAOYSA-N
Synonym erbium iii nitrate pentahydrate, erbium trinitrate pentahydrate, erbium nitrate pentahydrate, acmc-20al2v, er.3no3.5h2o, erbium nitrate-water 1/3/5, erbium iii nitrate pentahydrate, reacton, erbium nitrate, pentahydrate,
PubChem CID 53249207
SMILES O.O.O.O.O.[Er+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Pink
Physical Form Crystalline Powder and/or Crystals
Quantity 100 g
Packaging Plastic bottle
Assay Percent Range 33 to 39% (Er)
Linear Formula Er(NO3)3·5H2O
Solubility Information Solubility in water: soluble
IUPAC Name erbium(3+);trinitrate;pentahydrate
Formula Weight 443.35
Percent Purity 99.9%
Grade Trace Metal Basis
Chemical Name or Material Erbium(III) nitrate pentahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta

RUO – Research Use Only