missing translation for 'onlineSavingsMsg'
Learn More

Erbium ICP Standard, 1000 ppm Er in 3% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company PER1KN500

Packaging Natural Poly Bottle

 View more versions of this product

Catalog No. PER1KN500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

12061-16-4, 7697-37-2, 7732-18-5

Specifications

Erbium ICP Standard
12061-16-4 , 7697-37-2 , 7732-18-5
97.0, 4.81, 0.11
1mL = 1mg Er (1, 000ppm Er)Er2O3 in 3% HNO3
Natural Poly Bottle
93.0, 4.63, 0.11
Odorless
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 18 months

SDS
Documents

Documents