missing translation for 'onlineSavingsMsg'
Learn More

Erbium ICP Standard, 1000 ppm Er in 3% HNO3, Ricca Chemical

Catalog No. PER1KN100
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
PER1KN100 Natural LDPE Bottle
PER1KN500 Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PER1KN100 Supplier Ricca Chemical Company Supplier No. PER1KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12061-16-4 , 7697-37-2 , 7732-18-5

Specifications

Packaging Natural LDPE Bottle
CAS Min % 93.0, 4.63, 0.11
CAS Max % 97.0, 4.81, 0.11
Odor Odorless
Concentration or Composition (by Analyte or Components) 1mL = 1mg Er (1, 000ppm Er)Er2O3 in 3% HNO3
Chemical Name or Material Erbium ICP Standard
ShelfLife 18 months