missing translation for 'onlineSavingsMsg'
Learn More

epsilon-Caprolactam, 99%

Catalog No. AAL0699922
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0699922 100 g
AAL0699936 500 g
AAL069990E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0699922 Supplier Thermo Scientific Chemicals Supplier No. L0699922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 105-60-2 | C6H11NO | 113.16 g/mol

ε-Caprolactam is used as a monomer in the preparation of synthetic fibers of the polyamide as well as a solvent for high molecular weight polymers. It is also used in coatings and plasticizers. Further, it is used as a cross linking agent for polyurethanes. In addition to this, it is used to prepare nylon-6, which finds application in fibers and plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
ε-Caprolactam is used as a monomer in the preparation of synthetic fibers of the polyamide as well as a solvent for high molecular weight polymers. It is also used in coatings and plasticizers. Further, it is used as a cross linking agent for polyurethanes. In addition to this, it is used to prepare nylon-6, which finds application in fibers and plastics.

Solubility
Soluble in benzene, ethanol, water, chloroform, methanol, tetrahydrofurfuryl alcohol, ether, dimethylformamide, chlorinated hydrocarbons, cyclohexene and petroleum fractions.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 105-60-2
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00006936
InChI Key JBKVHLHDHHXQEQ-UHFFFAOYSA-N
Synonym epsilon-caprolactam, caprolactam, 6-caprolactam, 2-oxohexamethylenimine, aminocaproic lactam, 2-azacycloheptanone, hexahydro-2h-azepin-2-one, 2h-azepin-2-one, hexahydro, 6-hexanelactam, hexanolactam
PubChem CID 7768
ChEBI CHEBI:28579
IUPAC Name azepan-2-one
SMILES C1CCC(=O)NCC1

Specifications

Melting Point 68°C to 72°C
Density 1.01
pH 7 to 8.5
Boiling Point 270°C
Flash Point 152°C (305°F)
Quantity 100 g
Beilstein 106934
Merck Index 14,1761
Solubility Information Soluble in benzene,ethanol,water,chloroform,methanol,tetrahydrofurfuryl alcohol,ether,dimethylformamide,chlorinated hydrocarbons,cyclohexene and petroleum fractions.
Formula Weight 113.16
Percent Purity 99%
Chemical Name or Material epsilon-Caprolactam
Show More Show Less
Hazard Category H302+H312+H332-H315-H317-H319-H335-H500
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
EINECSNumber 203-313-2
RTECSNumber CM3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only