missing translation for 'onlineSavingsMsg'
Learn More

SOMNI Scientific™ EPS-3 Exposure Prevention System 3 Station Flowmeter Controlled Vacuum System

Catalog No. 04777167
Encompass
Change view
Click to view available options
Electrical Requirements:
110 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Electrical Requirements
04777167 110 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04777167 Supplier SOMNI Scientific™ Supplier No. WG15001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A self-contained exposure prevention system

  • 3 inlets allow scavenging of multiple devices simultaneously
  • Design includes a linear vacuum pump that produces up to 45 LPM of negative flow
  • It can be used where active vacuum is not available to safely remove anesthetic waste gas from multiple devices simultaneously
  • Prevents unnecessary exposure
  • Evacuate to SOMNI™ W.A.G. filter or any non-recirculating hood or vent appropriate for high volume
  • Energy saving timer, selectable up to 12 hours with auto power off
  • Lightweight, small footprint
  • Color coded for easy setup

Specifications

For Use With (Application) Animal Inhalant Anesthesia
For Use With (Equipment) AMD-17106, AMD-17004, TT-17000, TT-17001, TT-17002, WG-15003, MW-17005
Material Powder-coated Aluminum
Length (Metric) 30 cm
Voltage 110 V
Width (Metric) 20 cm
Electrical Requirements 110 V
Frequency 60 Hz
Includes Evacuation lines and outlet tubing with adapters

For Veterinary Use Only