Learn More
Eppendorf™ Reference™ 2 Multi-Channel Pipets- Trade-In Offer
Catalog No. 05412544
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
10 to 100 μL
30 to 300 μL
Number of Channels:
12
8
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Number of Channels
05412544 30 to 300 μL 12
05412539 0.5 to 10 μL 8
05412540 10 to 100 μL 8
05412542 0.5 to 10 μL 12
05412541 30 to 300 μL 8
05412543 10 to 100 μL 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 05412544 Supplier Eppendorf™ Supplier No. 2231302082
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Available in trade-in offer at reduced price
Exchange any brand pipet with brand new Reference 2 pipet (0.5 to 300μL) at low costs
  • One-button operation pipet offers fast and ergonomic handling with reduced operating effort and active aerosol reduction
  • Reduced repetitive strain injuries by not having to move your thumb sideways
  • Spring loaded tip cone improves user reproducibility and ergonomics by lowering tip attachment and ejection force
  • Secondary user adjustment supports easy adjustment of pipet for accurate pipetting without need for full calibration
  • Four digit volume display with magnifying window for easier volume identification, volume lock feature prevents accidental changes in volume
  • Clear haptic feedback is felt after blowout step which avoids accidental tip ejections
  • Embedded RFID chip contains all relevant pipet data and TrackIT software for simple identification and documentation
  • Offers high throughput option including spring-loaded nose cones which can be switched on/off optionally
  • High flexibility with ability to remove single cones
  • Ensures continuous identical pipet alignment during work process
  • Serial number is printed on multiple components of pipet
  • Volume labeled pipet for quick identification of volume size/tip size
  • Round upper part of pipet makes it possible to work in every position
  • Sturdy upper handle, movable parts and external edges made from stainless-steel make pipet robust
  • Quick connect lower part enables easy cleaning and maintenance of pipet
  • Fully autoclavable pipet with smooth surface and few rough edges
Specifications
Autoclavable Autoclavable
Number of Channels 12
Product Line Reference 2
Type Pipette
Volume (Metric) 30 to 300 μL