Learn More
Eppendorf™ Rotor for Centrifuges 5804 R and 5810 R
Eppendorf Rotor for Centrifuges 5804 R and 5810 R-precision meets power. Customize your centrifugation with varied capacities and speeds. Accessorize with versatile buckets and adaptors for efficient sample processing. Ideal for high-performance applications, ensuring accuracy and reproducibility.
Supplier:  Eppendorf™ 022637461
 Afficher plus de versions de ce produit
Numéro de catalogue. 361007279
Il en reste null
Description
Description
  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
  • Aerosol-tight options available.
Spécifications
Spécifications
Rotor
4,500 x g
Centrifuges 5804 R and 5810 R
A-4-44
8 x 50 mL
5,000 rpm
Buckets for 2 x 50 mL conical tubes (x2)
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS