Learn More
Electron Microscopy Sciences EPO-TEK H22 ADHESIVE 1OZ
Catalog No. 5033844
Encompass
Epo-Tek H22 is a two component, silver-filled epoxy system. Mixing ratio of silver resin paste and liquid hardener is 100:4.5. Pot life 16 hours, and shelf life is 6 months at room temperature. Smooth, free-flowing, slightly thixotropic paste. High Tg allows it to be used for high temperature applications (≤300 degree C). Contains no solvents – It is a NASA approved low outgasing epoxy. Excellent resistance to solvents, chemicals and moisture. Extended pot life and fast curing at low temperature <100 degree C. Designed for die bonding and sealing hybrid circuit. Recommended for SEM small angle cleavage and wafer bonding.
Catalog No. 5033844 Supplier Electron Microscopy Sciences Supplier No. 1267322
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Epo-Tek H22 is a two component, silver-filled epoxy system. Mixing ratio of silver resin paste and liquid hardener is 100:4.5. Pot life 16 hours, and shelf life is 6 months at room temperature. Smooth, free-flowing, slightly thixotropic paste. High Tg allows it to be used for high temperature applications (≤300 degree C). Contains no solvents – It is a NASA approved low outgasing epoxy. Excellent resistance to solvents, chemicals and moisture. Extended pot life and fast curing at low temperature <100 degree C. Designed for die bonding and sealing hybrid circuit. Recommended for SEM small angle cleavage and wafer bonding.