missing translation for 'onlineSavingsMsg'
Learn More

Epithiochlorohydrine, 97%

Catalog No. AC291390250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC291390250 25 g Glass bottle
AC291390050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC291390250 Supplier Thermo Scientific Chemicals Supplier No. 291390250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3221-15-6 | C3H5ClS | 108.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3221-15-6
Molecular Formula C3H5ClS
Molecular Weight (g/mol) 108.59
MDL Number MFCD00041199
InChI Key XRWMHJJHPQTTLQ-UHFFFAOYSA-N
Synonym 2-chloromethyl thiirane, thioepichlorohydrin, epithiochlorohydrin, thioepichlorhydrin, chloropropylene sulfide, chloromethyl thiirane, 3-chloropropylene sulfide, 2-thiiranylmethyl chloride, thiirane, chloromethyl, chloropropylene sulphide
PubChem CID 18588
IUPAC Name 2-(chloromethyl)thiirane
SMILES C1C(S1)CCl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2750g/mL
Boiling Point 69°C to 70°C (67.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5240 to 1.528
Quantity 25 g
Specific Gravity 1.275
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol,chloroform,diethylether
Formula Weight 108.59
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Epithiochlorohydrine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Causes serious eye damage.
Fatal if swallowed.
May cause an allergic skin reaction.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only