missing translation for 'onlineSavingsMsg'
Learn More

(-)-Epigallocatechin 98.0+%, TCI America™

Catalog No. E108450MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E108450MG 50 mg
E108410MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E108450MG Supplier TCI America Supplier No. E108450MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 970-74-1
Molecular Formula C15H14O7
Molecular Weight (g/mol) 306.27
MDL Number MFCD00075939
InChI Key XMOCLSLCDHWDHP-IUODEOHRSA-N
Synonym --epigallocatechin, epigallocatechin, epigallocatechol, l-epigallocatechin, epi-gallocatechin, antiscurvy factor c2, --epigallocatechol, l-epigallocatechol, --3,3',4',5,5',7-flavanhexol, 3,3',4',5,5',7-flavanhexol
PubChem CID 72277
ChEBI CHEBI:42255
IUPAC Name (2R,3R)-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)O

Specifications

Melting Point 210°C
Color White-Yellow
Quantity 50 mg
Formula Weight 306.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Epigallocatechin
RTECSNumber KB5100000
TSCA No
Recommended Storage Refrigerator