missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (-)-Epigallocatechin gallate, 95%

Catalog No. AC449010100
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC449010100 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC449010100 Supplier Thermo Scientific Chemicals Supplier No. 449010100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 989-51-5 | C22H18O11 | 458.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 989-51-5
Molecular Formula C22H18O11
Molecular Weight (g/mol) 458.37
InChI Key WMBWREPUVVBILR-WIYYLYMNSA-N
Synonym =--epigallocatechin gallate, egcg, epigallocatechin gallate, epigallocatechin 3-gallate, tea catechin, epigallocatechin-3-gallate, =--epigallocatechin-3-o-gallate, teavigo, =--epigallocatechol gallate, epigallocatechin-3-monogallate
PubChem CID 65064
ChEBI CHEBI:4806
IUPAC Name [(2R,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 10 mg
Merck Index 153573
Solubility Information Solubility in water: partially soluble. Other solubilities: practically insoluble in acetone,dmso,methanol
Formula Weight 458.37
Percent Purity 95%
Chemical Name or Material (-)-Epigallocatechin gallate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only