missing translation for 'onlineSavingsMsg'
Learn More

(-)-Epigallocatechin, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015985
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015985 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015985 Supplier Merck Emd Millipore Supplier No. 081081MGF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

(−)-Epigallocatechin (EGC) belongs to the tea catechins group of polyphenols.

Featured Industries: Food and Beverages

Specifications

CAS 970-74-1
Linear Formula C15H14O7
MDL Number MFCD00075939
Synonym (-)-cis-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2 H)-benzopyran-3,5,7-triol; (-)-cis-3,3′,4′,5,5′,7-Hexahydroxyflavane
Type Analytical Standard
Quantity 1 mg
Formula Weight 306.27
Percent Purity ≥90% (HPLC)
Grade Analytical Standard