missing translation for 'onlineSavingsMsg'
Learn More

Epichlorohydrin 99.0+%, TCI America™

Catalog No. E0012500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0012500G 500 g
E001225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0012500G Supplier TCI America Supplier No. E0012500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106-89-8
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.522
MDL Number MFCD00005132
InChI Key BRLQWZUYTZBJKN-UHFFFAOYSA-N
Synonym epichlorohydrin, 2-chloromethyl oxirane, epichlorhydrin, 1-chloro-2,3-epoxypropane, glycidyl chloride, oxirane, chloromethyl, epichlorhydrine, 1,2-epoxy-3-chloropropane, 2,3-epoxypropyl chloride, chloropropylene oxide
PubChem CID 7835
ChEBI CHEBI:37144
IUPAC Name 2-(chloromethyl)oxirane
SMILES C1C(O1)CCl

Specifications

Melting Point -26°C
Color Colorless
Boiling Point 117°C
Quantity 500 g
UN Number 2023
Formula Weight 92.52
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Epichlorohydrin
EINECSNumber (2)-0275
RTECSNumber TX4900000
TSCA Yes