missing translation for 'onlineSavingsMsg'
Learn More

(±)-Epichlorohydrin, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020350
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020350 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020350 Supplier Merck Emd Millipore Supplier No. 025781ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Epichlorohydrin (ECH) is a colorless, volatile and highly reactive organic compound which is utilized as a precursor in the production of glycerol, plastics, drugs and paints.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 106-89-8
Ignition Point 779°F
Vapor Pressure 13.8 mmHg (21.1°C)
Vapor Density 3.2 (vs. air)
Density 1.183 g/mL (at 25°C)
Flash Point 32°C - closed cup
Linear Formula C3H5 ClO
MDL Number MFCD00005132
Melting Point -57°C
Boiling Point 115°C to 117°C
Synonym (+/-)-2-(Chloromethyl)oxirane; 1-Chloro-2,3-epoxypropane
Type Analytical Standard
Quantity 1 mL
Formula Weight 92.52
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less