missing translation for 'onlineSavingsMsg'
Learn More

EPA 8270 LCS Mix 1 100 μg/mL each component in acetone: methylene chloride (9:1), MilliporeSigma™ Supelco™

Catalog No. 111016972
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016972 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016972 Supplier Merck Emd Millipore Supplier No. 46853U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

EPA 8270 LCS Mix 1 is utilized for identifying and quantifying semivolatile organic compounds on the EPA's Appendix VIII and Appendix IX lists in solid wastes and groundwater at active hazardous treatment, storage, and disposal sites. The pre-requisite for EPA 8270 method is the use of gas chromatography with a selective detector or gas chromatography/mass spectrometry. Featured Industries: Environmental

Specifications

Packaging Ampule
CAS 67-64-1
Flash Point -19°C
Concentration 100 μg/mL each component in acetone: methylene chloride (9:1)
Type Organic Pollutant Standard
Quantity 25 mL
Grade Analytical Standard