missing translation for 'onlineSavingsMsg'
Learn More

EPA 8270 GC-MS Tuning Solution, Certified Reference Material, 50 μg/mL Each Component in Dichloromethane, MilliporeSigma™ Supelco™

Catalog No. 111016434
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016434 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016434 Supplier Merck Emd Millipore Supplier No. 47387

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

Packaging Ampule
Product Line TraceCERT
Concentration 50 μg/mL each component in dichloromethane
Quantity 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) Benzidine; 4,4 '-DDT; Decafluorotriphenylphosphine; Pentachlorophenol

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC