missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA 552 Halogenated Acetic Acids Mix, Certified Reference Material, 2,000 μg/mL, MilliporeSigma™ Supelco™
Description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| Concentration | 2,000 μg/mL each component in methyl tert -butyl ether |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | Bromoacetic acid; Bromochloroacetic acid; Chloroacetic acid; Dibromoacetic acid; Dichloroacetic acid; Trichloroacetic acid |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC