missing translation for 'onlineSavingsMsg'
Learn More

EP Reference Solution BY7, Brownish-Yellow Reference Solution, Ricca Chemical

Catalog No. 28774
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28774 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 28774 Supplier Ricca Chemical Company Supplier No. 28774

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Brownish-Yellow Reference Solution
Packaging Amber Glass Bottle
CAS 10025-77-1, 7647-01-0, 7732-18-5, 7758-99-8, 7791-13-1
Physical Form Liquid
Color Brown or Green/Yellow
pH Acidic
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Miscible with water
Solution Type Calibration Solution
Type Color Reference Standard
Quantity 120 mL
Grade Laboratory
Show More Show Less