Complete arsenic limit test distillation apparatus as described in European Pharmacopeia, 1997, page 51. Includes a 125ml conical flask w/ 24/40 joint, a capillary chimney w/ a pulled tip, and an adapter tube w/ #5 o-ring joint with hooks.
Numéro de catalogue.
NC9596429
Fournisseur
Ace GlassCode fournisseur
654446
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Complete arsenic limit test distillation apparatus as described in European Pharmacopeia, 1997, page 51. Includes a 125ml conical flask w/ 24/40 joint, a capillary chimney w/ a pulled tip, and an adapter tube w/ #5 o-ring joint with hooks.