missing translation for 'onlineSavingsMsg'
Learn More

Eosin Y TS, (U.S.P. Test Solution), Spectrum™ Chemical

Catalog No. 18602414
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602414 1 L Amber Glass Bottle
18602413 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18602414 Supplier Spectrum Chemical Mfg Cor Supplier No. E1461LT
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

E-146, 15086-94-9

Spectrum™ Chemical Eosin Y TS, (U.S.P. Test Solution) is a solution, sometimes designated with 'TS or Test Solution', that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

Chemical Identifiers

CAS 17372-87-1 , 7732-18-5
Molecular Formula C20H6Br4Na2O5
Molecular Weight (g/mol) 691.86
InChI Key SEACYXSIPDVVMV-UHFFFAOYSA-L
IUPAC Name disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12

Specifications

CAS Max % 0.1%, 99.9%
Grade Chemical Solution
Packaging Amber Glass Bottle
Quantity 1 L
Chemical Name or Material Eosin Y TS