missing translation for 'onlineSavingsMsg'
Learn More

Eosin Y TS, 0.5% (w/v), Adsorption Indicator for Argentometric Titrations, Ricca Chemical

Catalog No. 28584
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
28584 120 mL Natural Poly Bottle
285816 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 28584 Supplier Ricca Chemical Company Supplier No. 28584

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Eosin Y
Name Note Test Solution, Adsorption Indicator For Argentometric Titrations
CAS 17372-87-1, 7732-18-5
Concentration 0.5% (w/v)
Packaging Natural Poly Bottle
Molecular Formula C20H12Br4Na2O8
Solvent or Matrix Aqueous Solution
Color Dark Red
Quantity 120 mL
InChI Key MASXMTNVNASWNH-UHFFFAOYSA-L
Physical Form Liquid
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
IUPAC Name disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
Molecular Weight (g/mol) Mixture
PubChem CID 91886399
Grade Laboratory
Show More Show Less