missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma-Aldrich Eosin Y Solution, Aqueous
Description
Certified Eosin Y, 0.5% (w/v) in water. Not acidified.
- Form: solution
- Shelf life: expiry date on the label
- IVD: for in vitro diagnostic use
- Concentration: 0.5% (w/v) in water
- Applications: hematology and histology
- Storage temperature: room temperature
Specifications
Specifications
| Quantity | 1 L |
| Product Type | Eosin Y |