missing translation for 'onlineSavingsMsg'
Learn More

Eosin Y Disodium Salt, Spectrum™ Chemical

Catalog No. 18610365
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610365 5 kg Amber Glass Bottle
18610363 25 g Amber Glass Bottle
18610362 100 g Amber Glass Bottle
18610361 1 kg Amber Glass Bottle
18610364 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18610365 Supplier Spectrum Chemical Mfg Cor Supplier No. EO1105KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17372-87-1
Molecular Weight (g/mol) 691.85

Specifications

Physical Form Solid
CAS Max % 1
Packaging Amber Glass Bottle
Color Red
Quantity 5 kg
Grade Ungraded
Chemical Name or Material Eosin Y Disodium Salt

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.