missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Entacapone

Catalog No. 472010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
472010 10 mg
472050 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 472010 Supplier Tocris Bioscience™ Supplier No. 4720/10

Potent COMT inhibitor; blocks α-synuclein aggregation

Potent catechol O-methyltransferase (COMT) inhibitor (IC50 values are 14.3, 20.1 and 73.3 nM for rat liver soluble COMT, total COMT and membrane-bound COMT respectively). Increases bioavailability of levodopa when given as an adjunct therapy for Parkinson's disease. Inhibits α-synuclein aggregation in an in vitro assay; blocks α-synuclein-induced cell death in PC-12 cells.

Chemical Identifiers

CAS 130929-57-6
Molecular Formula C14H15N3O5

Specifications

Quantity 10 mg
Formula Weight 305.29
Purity >99%
Molecular Weight (g/mol) 305.29
Chemical Name or Material Entacapone
Recommended Storage Store at Room Temperature