accessibility menu, dialog, popup

UserName

CAS RN 130929-57-6

IUPAC Name: (2E)-2-cyano-3-(3,4-dihydroxy-5-nitrophenyl)-N,N-diethylprop-2-enamide
Synonyms: entacapone comtess entacapona entacapona [inn-spanish] 恩他卡朋 entacaponum comtan
Molecular Weight (g/mol): 305.29
Molecular Formula: C14H15N3O5
InChi Key: JRURYQJSLYLRLN-BJMVGYQFSA-N
SMILES: CCN(CC)C(=O)C(=C\C1=CC(O)=C(O)C(=C1)[N+]([O-])=O)\C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)