missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Entacapone

Catalog No. AC466220010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466220010 1 g
AC466222500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466220010 Supplier Thermo Scientific Chemicals Supplier No. 466220010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 130929-57-6
Molecular Formula C14H15N3O5

Specifications

Quantity 1 g
Formula Weight 305.29
InChI Key JRURYQJSLYLRLN-BJMVGYQFSA-N
SMILES CCN(CC)C(=O)C(=C\C1=CC(O)=C(O)C(=C1)[N+]([O-])=O)\C#N
IUPAC Name (2E)-2-cyano-3-(3,4-dihydroxy-5-nitrophenyl)-N,N-diethylprop-2-enamide
Molecular Weight (g/mol) 305.29
Chemical Name or Material Entacapone

RUO – Research Use Only