missing translation for 'onlineSavingsMsg'
Learn More

Ensulizole(2-Phenylbenzimidazole 5-sulfonic acid) Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013795
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013795 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013795 Supplier Merck Emd Millipore Supplier No. PHR13011G

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 27503-81-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C13H10N2O3 S
MDL Number MFCD00053007
Melting Point >300°C
Synonym 2-Phenyl-5-benzimidazolesulfonic acid; Ensulizole
Type Certified Reference Material
Quantity 1 g
Formula Weight 274.30
Grade Certified Reference Material
Show More Show Less