missing translation for 'onlineSavingsMsg'
Learn More
Learn More
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
18048036
![]() |
N/A | 1 kg |
Each for $1,996.51
|
|
|||||
18048050
![]() |
N/A | 100 g |
Each for $555.10
|
|
|||||
18048051
![]() |
N/A | 10 g |
Each for $85.88
|
|
|||||
18048052
![]() |
N/A | 25 g |
Each for $187.07
|
|
|||||
18048093
![]() |
N/A | 500 g |
Each for $1,199.13
|
|
|||||
Description
Enrofloxacin USP is a pale yellow to light yellow crystalline powder.
Chemical Identifiers
| 93106-60-6 |
Specifications
| α-Carboxybenzyl Penicillin Disodium Salt | |
| 93106-60-6 | |
| 98.5 to 101.5% |
| Crystalline Powder | |
| Light Yellow to Yellow | |
| USP |
