missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enrofloxacin USP, Medisca™

Supplier: Medisca Pharmaceutique Inc 257404*
| Quantity | 25 g |
|---|
Description
Enrofloxacin USP is a pale yellow to light yellow crystalline powder.
Chemical Identifiers
| 93106-60-6 |
Specifications
| α-Carboxybenzyl Penicillin Disodium Salt | |
| 93106-60-6 | |
| 25 g | |
| USP |
| Crystalline Powder | |
| Light Yellow to Yellow | |
| 98.5 to 101.5% |