missing translation for 'onlineSavingsMsg'
Learn More

Enrofloxacin USP, Medisca™
GREENER_CHOICE

Catalog No. 18048093
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18048093 500 g
18048036 1 kg
18048050 100 g
18048051 10 g
18048052 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18048093 Supplier Medisca Pharmaceutique Inc Supplier No. 257408
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Enrofloxacin USP is a pale yellow to light yellow crystalline powder.

Chemical Identifiers

CAS 93106-60-6

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Crystalline Powder
Color Light Yellow to Yellow
Quantity 500 g
Percent Purity 98.5 to 101.5%
Grade USP