missing translation for 'onlineSavingsMsg'
Learn More

CEP ENPAC™ Spillpal Drum Pallets

Catalog No. 19048806
Change view
Click to view available options
Dimensions (L x W x H):
2 x 2 x 4 in.
2 x 4 x 4 in.
4 x 4 x 4 in.
4 x 6 x 4 in.
4 x 8 x 4 in.
Material:
Nylon
Polyethylene
Type:
1-drum Spill Pallet
2-drum Spill Pallet
4-drum Spill Pallet
6-drum Spill Pallet
8-drum Spill Pallet
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H) Material Type
19048806 4 x 8 x 4 in. Nylon 8-drum Spill Pallet
19048800 2 x 2 x 4 in. Polyethylene 1-drum Spill Pallet
19048802 2 x 4 x 4 in. Polyethylene 2-drum Spill Pallet
19048803 4 x 4 x 4 in. Nylon 4-drum Spill Pallet
19048805 4 x 6 x 4 in. Nylon 6-drum Spill Pallet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19048806 Supplier CEP Supplier No. 5775 YE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides incidental spill control during temporary storage of drums, machine parts, tanks and generators

  • Flexible four inch foam sidewalls spring back to form, allowing easy drive-in and drive-out with a hand truck or forklift
  • Designed for 37.8L (10 gal.) drums
  • Able to be folded and stored
  • Reusable

Specifications

Certifications/Compliance EPA, 40 CFR 264.175
Color Yellow
Capacity (English) 80 gal.
Material Nylon
Type 8-drum Spill Pallet
For Use With (Application) Used to store hazardous material
Holds 8 Drums
Includes Foam sidewalls
Dimensions (L x W x H) 4 x 8 x 4 in.
Length (English) 4 ft.
Length (Metric) 1.21 m
Width (English) 8 ft.
Width (Metric) 2.43 m
Height (English) 4 in.
Height (Metric) 10.16 cm
Weight (English) 7.5 lb.
Weight (Metric) 3.4 kg
Show More Show Less