Learn More
LGC Genomics Llc (Lucigen) Endura Electrocompetent Cells, 12 Rxns (DUOs)Clone and maintain unstable or repetitive sequences and lentiviral libraries with high efficiency. Ideal for GeCKO CRISPR library transformation.
Numéro de catalogue. 501047945
Encompass
Lucigens Endura Competent Cells are a commonly used strain for cloning sequences that suffer unwanted recombination events in other strains. Clones with inverted repeats or other sequences prone to recombination are commonly found in retroviral genes, and require cells such as Endura to be propagated stably.
Numéro de catalogue. 501047945 Fournisseur LGC Genomics Llc Code fournisseur 602421
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Lucigens Endura Competent Cells are a commonly used strain for cloning sequences that suffer unwanted recombination events in other strains. Clones with inverted repeats or other sequences prone to recombination are commonly found in retroviral genes, and require cells such as Endura to be propagated stably.