missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endothal -WS, MilliporeSigma™ Supelco™
Description
Featured Industries: Environmental
Specifications
Specifications
| CAS | 67-56-1 |
| Flash Point | 11°C - closed cup - Solvent |
| Type | Proficiency Testing Material |
| Quantity | 2 mL |