missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ endo-IWR 1

Catalog No. 353210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
353210 10 mg
353250 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 353210 Supplier Tocris Bioscience™ Supplier No. 3532/10*

Wnt/β-catenin signaling inhibitor; axin stabilizer

endo-IWR 1 is an inhibitor of Wnt signaling. Induces an increase in axin2 protein levels; promotes β-catenin phosphorylation by stabilizing Axin-scaffolded destruction complexes. Promotes endothelial cell specification of cardiac progenitor cells

Negative Control also available.

For more information about how endo-IWR 1 may be used, see our protocol: Generation and Propagation of EPS cells (LCDM Cocktail) .

Chemical Identifiers

CAS 1127442-82-3
Molecular Formula C25H19N3O3

Specifications

Quantity 10 mg
Formula Weight 409.44
Purity >98%
Molecular Weight (g/mol) 409.445
Chemical Name or Material endo-IWR 1
Recommended Storage Store at Room Temperature