accessibility menu, dialog, popup

UserName

ONOONHN

CAS RN 1127442-82-3

IUPAC Name: 4-{3,5-dioxo-4-azatricyclo[5.2.1.0²,⁶]dec-8-en-4-yl}-N-(quinolin-8-yl)benzamide
Synonyms: 4-{3,5-dioxo-4-azatricyclo[5.2.1.0²,⁶]dec-8-en-4-yl}-N-(quinolin-8-yl)benzamide
Molecular Weight (g/mol): 409.45
Molecular Formula: C25H19N3O3
InChi Key: ZGSXEXBYLJIOGF-UHFFFAOYNA-N
SMILES: O=C(NC1=C2N=CC=CC2=CC=C1)C1=CC=C(C=C1)N1C(=O)C2C3CC(C=C3)C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results