Learn More
New England Biolab Ltd Endo F3 240 units
Numéro de catalogue. NC1309545
Encompass
Endo F3 is an endoglycosidase that cleaves within the chitobiose core of N-linked fucosylated-biantennary and triantennary complex oligosaccharides from glycoproteins. Endo F3 is tagged with chitin binding domain (CBD) for easy removal from a reaction.
Numéro de catalogue. NC1309545 Fournisseur New England Biolab Ltd Code fournisseur P0771S
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Endo F3 is an endoglycosidase that cleaves within the chitobiose core of N-linked fucosylated-biantennary and triantennary complex oligosaccharides from glycoproteins. Endo F3 is tagged with chitin binding domain (CBD) for easy removal from a reaction.