missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enalaprilat, ≥98,0% (HPLC), MilliporeSigma™ Supelco™
Spécifications
Spécifications
| CAS | 76420-72-9 |
| Physical Form | Sympa |
| Linear Formula | C18H24N2O5 |
| MDL Number | MFCD00865786 |
| Synonym | (2 S)-1-[(2 S)-2-[[(1 S)-1-Carboxy-3-phenyl-propyl]amino]propanoyl]pyrrolidine-2-carboxylic acid; N-[(1 S)-1-Carboxy-3-phenylpropyl]-L-alanyl-L-proline |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 348.39 |
| Percent Purity | ≥98.0% (HPLC) |
| Grade | Norme analytique |