missing translation for 'onlineSavingsMsg'
Learn More

EN 14105/D6584 Monoglyceride Stock Solution, 10 mg/mL Each Component in Pyridine, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111017146
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017146 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017146 Supplier Merck Emd Millipore Supplier No. 49446U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sigma Life Science is committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention. For more information see the article in biofiles.

Specifications

Concentration 10 mg/mL each component in pyrid ine
Synonym Biodiesel impurity; Commercial Mixture of Monoglycerides; Monoglyceride mix; Monoglyceride mixture; Monoglyceride standard; Monoglycerides
Quantity 1 mL
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 1-Monooleoyl-rac-glycerol; 1-Monostearoyl-rac-glycerol; 1-Monopalmitoyl-rac-glycerol