missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Emtricitabine

Catalog No. AC462070010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462070010 1 g
AC462070050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462070010 Supplier Thermo Scientific Chemicals Supplier No. 462070010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 143491-57-0 | C8H10FN3O3S

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 143491-57-0
Molecular Formula C8H10FN3O3S

Specifications

Quantity 1 g
Solubility Solubility in water: soluble. Other solubilities: very soluble in methanol,very slightly soluble in acetonitrile
Merck Index 153622
Melting Point 136°C to 140°C
Formula Weight 247.24
Chemical Name or Material Emtricitabine

RUO – Research Use Only