Learn More
MilliporeSigma™ Supelco™ Empty Glass GC Column, for Agilent™ 5880, 5890, 6890, 7820, and 7890 (Configuration A)
Empty column may be used for packing adsorbents for temperature-programmed desorption (TPD) instrumentation.
Supplier:  MilliporeSigma™ Supelco™ 21641
 View more versions of this product
Catalog No. 111004386
Only null left
Explore available promotions
Description
Description
Silane treated empty glass column. Fittings are not included. This column fits Agilent 5880, 5890, 6890, 7820, and 7890 (configuration A, on-column injection, all detectors except TCD).
Specifications
Specifications
2 mm
Gas Chromatography (GC)
Glass Column
Empty GC
1.8 m
None
Product Suggestions
Product Suggestions