missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDS Empore™ SDB-RPS Extraction Disks
Formerly 3M™ Empore™ SDB-RPS Solid Phase Extraction Disks (SPE): 2241 & 2341. Reduce extraction time and solvent use for analytical preparation of aqueous environmental samples.
Supplier: CDS 98060402272EA
Description
- Approved for use in EPA Method 515.2
- Used to collect the more water-soluble analytes from water samples
- Adsorbent is effective for explosives, pesticides, phenolics, and drugs of abuse
- Provide up to 90% solvent reduction, excellent processing speed, and contamination-free extractions with accurate and reproducible results
- Available with a variety of packings in 47 and 90mm sizes
Specifications
| EPA Method 515.2 | |
| Analytical Preparation of Aqueous Environmental Samples | |
| Extraction Disk | |
| 90% | |
| 9.0 cm |
| Reverse | |
| SDB-RPS | |
| 30/Cs. | |
| 9.0 cm |